Learn More
Chemglass Life Sciences 250mL Addition Funnel, 24/40 Joint Size, 2mm Stpk, 375mm OAH
SDP
Catalog No. CG170803
Encompass_Preferred
250mL Addition Funnel, 24/40 Joint Size, 2mm Stopcock, 375mm OAH. Cylindrical shape addition funnel with PTFE stopcocks. For replacement stoppers see CG-3000.
Catalog No. CG170803 Supplier Chemglass Life Sciences Supplier No. CG170803
250mL Addition Funnel, 24/40 Joint Size, 2mm Stopcock, 375mm OAH. Cylindrical shape addition funnel with PTFE stopcocks. For replacement stoppers see CG-3000.