Learn More
Chemglass Life Sciences 250mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Stpk, 350mm OAH
SDP
Catalog No. CG171222
Encompass_Preferred
250mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Stopcock, 350mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171222 Supplier Chemglass Life Sciences Supplier No. CG171222
250mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Stopcock, 350mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.