Learn More
Ace Glass, Inc. Mantle, 250mL, for 83mm diameter spherical or cylindrical flask, 84mm ID x 41.4mm height, 180w-115v, CSA rated
SDP
Catalog No. 50977323
Encompass
250mL aluminum housed, table-model mantle for 83mm diameter spherical or cylindrical flask. 84mm I.D. x 41.4mm height. Maximum internal operating temperature is 450 Deg. centigrade. 180w-115v, cULus rated. Power cord included.
Catalog No. 50-977-323 Supplier Ace Glass, Inc. Supplier No. 1205313
250mL aluminum housed, table-model mantle for 83mm diameter spherical or cylindrical flask. 84mm I.D. x 41.4mm height. Maximum internal operating temperature is 450 Deg. centigrade. 180w-115v, cULus rated. Power cord included.