Learn More
Ace Glass, Inc. Beaker, 250mL, 68mm OD, 90mm height, cs/48, sp/12, 1000-250graduation range 25-200mL, graduation interval 25mL
SDP
Catalog No. 509557012
Encompass
250mL PYREX brand beaker, with lip, pouring spout, and double scale graduation. Dimensions: 68mm O.D. x 90mm H. Graduation range: 25-200mL. Graduation interval is 25mL. Case/48.
Catalog No. 50-955-7012 Supplier Ace Glass, Inc. Supplier No. 532821
Add to Cart
Add to Cart
250mL PYREX brand beaker, with lip, pouring spout, and double scale graduation. Dimensions: 68mm O.D. x 90mm H. Graduation range: 25-200mL. Graduation interval is 25mL. Case/48.