Learn More
Ace Glass, Inc. Beaker, 250mL, jacketed, glass hose connections for 3/8intubing, 89mm inside height, 65mm inside diameter
SDP
Catalog No. 509557022
Encompass_Preferred
250mL jacketed beaker with one upper and one lower glass hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 89mm I.H. x 65mm I.D. Beaker does not have a pouring spout.
Catalog No. 50-955-7022 Supplier Ace Glass, Inc. Supplier No. 534005
250mL jacketed beaker with one upper and one lower glass hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 89mm I.H. x 65mm I.D. Beaker does not have a pouring spout.