Learn More
Waters Corp Bottle in Rack Domino, 250 mL, 18-Bottle Capacity, 3x2 Format
Catalog No. NC3597615 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 250mL Bottle in 18x Rack Domino is a specialized consumable holder for automated liquid handling with the Andrew+ pipetting robot. It accommodates a rack of up to 18 large bottles and occupies a 3x2 format on the robot workbench, equivalent to six standard Dominos. This machine-readable Domino facilitates seamless automation with QR and ARUCO codes for labware info and spatial positioning, crucial for integrating various labware into automation setups.
  • Holds a large format rack with up to 18 large bottles
  • 3x2 format, occupying 3 horizontal and 2 vertical positions on the robot workbench
  • Equivalent to the space taken by 6 standard Dominos
  • Machine-readable consumable holder for various consumable types
  • Equipped with QR codes for critical labware information
  • Includes ARUCO codes for spatial positioning
  • Indispensable component for automation configurations
Catalog No. NC3597615 Supplier Waters Corp Supplier No. 186010515
Add to Cart
Add to Cart
Request a Quote
The 250mL Bottle in 18x Rack Domino is a specialized consumable holder for automated liquid handling with the Andrew+ pipetting robot. It accommodates a rack of up to 18 large bottles and occupies a 3x2 format on the robot workbench, equivalent to six standard Dominos. This machine-readable Domino facilitates seamless automation with QR and ARUCO codes for labware info and spatial positioning, crucial for integrating various labware into automation setups.
  • Holds a large format rack with up to 18 large bottles
  • 3x2 format, occupying 3 horizontal and 2 vertical positions on the robot workbench
  • Equivalent to the space taken by 6 standard Dominos
  • Machine-readable consumable holder for various consumable types
  • Equipped with QR codes for critical labware information
  • Includes ARUCO codes for spatial positioning
  • Indispensable component for automation configurations