Learn More
Ace Glass, Inc. 250mL gas wash bottle only. Use with 7162, 7163 & 7164stoppers
SDP
Catalog No. 50974102
Encompass_Preferred
250mL gas washing bottle, to be used with gas washing stoppers of different porosities.
Catalog No. 50-974-102 Supplier Ace Glass, Inc. Supplier No. 716252
250mL gas washing bottle, to be used with gas washing stoppers of different porosities.