Learn More
Chemglass Life Sciences Bottle Only, 250mL Gas Washing
SDP
Catalog No. 501531009
Encompass_Preferred
Bottle Only, 250mL Gas Washing. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion. Image shown is the complete bottle.
Catalog No. 50-153-1009 Supplier Chemglass Life Sciences Supplier No. CG111207
Bottle Only, 250mL Gas Washing. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion. Image shown is the complete bottle.