Learn More
CELLTREAT Scientific Products 250mL Bottle Top Filter, 0.22m PES Filter, 75mm Diameter, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229716
Encompass_Preferred
CELLTREAT Scientific Products 250mL 0.22m PES Bottle Top Filter. Easy-to-read molded graduations marks on bottle top filter. Bold easy-to-read printed membrane and pore size for easy identification. Curved bottle design for easy handling. Printed lot number and expiration date on every bottle top. Improved filter support provides faster flow rates. Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping. Polyethersulfone (PES) membrane is for high flow and low binding. 0.22m PES membrane is the first choice for sterile filtration of cell culture media. Bottle top filters fit on CELLTREAT solution bottles or other standard 45mm threaded solution storage bottles. Bottle top filters do not include solution bottles. Membrane diameter 75mm. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 24/case.
Catalog No. 50-202-052