Learn More
NEST SCIENTIFIC INC 250mL Centrifuge Tube with Bi-directional Transfer Cap with Silicon Tube (50cm 1/8" ID 1/4" OD), Male Luer Lock Connector with Luer Cap, Vent Filter (0.22 μm 24mm), Sterile, 1/pk, 4/cs - C60922-EFB050A
Catalog No. NC3311157
Encompass_Preferred
250mL Centrifuge Tube with Bi-directional Transfer Cap with Silicon Tube (50cm 1/8" ID 1/4" OD), Male Luer Lock Connector with Luer Cap, Vent Filter (0.22 μm Φ24mm), Sterile, 1/pk, 4/cs
Catalog No. NC3311157 Supplier NEST SCIENTIFIC INC Supplier No. C60922EFB050A
250mL Centrifuge Tube with Bi-directional Transfer Cap with Silicon Tube (50cm 1/8" ID 1/4" OD), Male Luer Lock Connector with Luer Cap, Vent Filter (0.22 μm Φ24mm), Sterile, 1/pk, 4/cs