Learn More
Bellco Glass, Inc. 250mL Centrifuge Bottle
SDP
Supplier:  Bellco Glass, Inc. 229467000
Encompass_Preferred
DescriptionCentrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samplesManufactured from high quality polypropylene for excellent chemical resistanceBottle must be filled 80% of total capacity to obtain maximum resultsCentrifuge Bottles are reusable and autoclavablePackaged non-sterile, 2 per bag Pack Description2/BagCase Quantity2SterileNoAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume250mLGraduatedNoMax Force (RCF)g27500
Catalog No. 50-232-9313