Learn More
CELLTREAT Scientific Products 250mL Centrifuge Bottle (Polypropylene), Non-sterile
SDP
Catalog No. 50828733
Encompass_Preferred
CELLTREAT Scientific Products 250mL Centrifuge Bottles. Centrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samples. Manufactured from high quality polypropylene for excellent chemical resistance. Bottle must be filled 75% of total capacity to obtain maximum results. Centrifuge Bottles are reusable and autoclavable. Max force (RCF) g 27500. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 2 per re-sealable bag.
Catalog No. 50-828-733 Supplier CELLTREAT Scientific Products Supplier No. 229467
CELLTREAT Scientific Products 250mL Centrifuge Bottles. Centrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samples. Manufactured from high quality polypropylene for excellent chemical resistance. Bottle must be filled 75% of total capacity to obtain maximum results. Centrifuge Bottles are reusable and autoclavable. Max force (RCF) g 27500. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 2 per re-sealable bag.