Learn More
CELLTREAT Scientific Products 250mL Conical Bottom Centrifuge Tube, Sterile
SDP
Catalog No. 501117985
Encompass_Preferred
CELLTREAT Scientific Products 250mL Centrifuge Tube. For applications requiring large volume centrifugation. Tubes include molded graduation lines. Polyethylene (HDPE) cap with integral plug seal provides a secure closure. To suport tube during centrifugation, use of a rotor cushion is required. Temperature range of -80¿C to 120¿C. Max force (RCF) g 7500. Every case is printed with the lot number for quality traceability. Packaged sterile, 6/re-sealable bag, 48/case.
Catalog No. 50-111-7985 Supplier CELLTREAT Scientific Products Supplier No. 229466
CELLTREAT Scientific Products 250mL Centrifuge Tube. For applications requiring large volume centrifugation. Tubes include molded graduation lines. Polyethylene (HDPE) cap with integral plug seal provides a secure closure. To suport tube during centrifugation, use of a rotor cushion is required. Temperature range of -80¿C to 120¿C. Max force (RCF) g 7500. Every case is printed with the lot number for quality traceability. Packaged sterile, 6/re-sealable bag, 48/case.