Learn More
Consolidated Plastics 250 mL Polypropylene Disposable BeakerQuantity per case: 10 packages of 50
SDP
Catalog No. NC1092975
Encompass
Polypropylene Plastic BeakersTranslucent plastic beakers are resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents. Autoclavable beakers have excellent temperature resistance (250oF). The beakers are molded with a special "dripless" lip and have excellent clarity.
Catalog No. NC1092975 Supplier Consolidated Plastics Supplier No. 45383
Polypropylene Plastic BeakersTranslucent plastic beakers are resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents. Autoclavable beakers have excellent temperature resistance (250oF). The beakers are molded with a special "dripless" lip and have excellent clarity.