Learn More
Thomas Scientific 250mL Filter System, PES Filter, 0.22æm, 50mm, Sterile
SDP
Supplier:  Thomas Scientific 1159T64
Encompass_Preferred
12/cs Angled hose barb adapter fits 3 size hose diameters and improves stability Polyethersulfone (PES) membrane for high flow and low binding Two membrane pore sizes: 0.22Ým or 0.45Ým Filter systems include bottle top filter and solution bottle with cap Bottle top filters fit standard 45mm threaded solution storage bottles Packaged sterile (gamma irradiated) in bags
Catalog No. 50-145-9561