Learn More
Ace Glass, Inc. 250mL flask 1/4" hose connection
SDP
Catalog No. 50973979
Encompass_Preferred
250mL heavy wall filter flask with a tooled neck for uniform stopper fit. Side hose connection is a #11 Ace-Thred that accepts a PTFE tubing connector with a silicone o-ring and nylon bushing to make an easier, safer, more convenient vacuum hose connection. Includes flask, 1/4 tubing connector, and nylon bushing.
Catalog No. 50-973-979 Supplier Ace Glass, Inc. Supplier No. 698347
250mL heavy wall filter flask with a tooled neck for uniform stopper fit. Side hose connection is a #11 Ace-Thred that accepts a PTFE tubing connector with a silicone o-ring and nylon bushing to make an easier, safer, more convenient vacuum hose connection. Includes flask, 1/4 tubing connector, and nylon bushing.