Learn More
Ace Glass, Inc. 250mL flask 3/8" hose connection
SDP
Catalog No. 50973976
Encompass_Preferred
250mL heavy wall filter flask with a tooled neck for uniform stopper fit. Side hose connection is a #11 Ace-Thred that accepts a PTFE tubing connector with a silicone o-ring and nylon bushing to make an easier, safer, more convenient vacuum hose connection. Includes flask, 3/8 tubing connector, and nylon bushing.
Catalog No. 50-973-976 Supplier Ace Glass, Inc. Supplier No. 698328
250mL heavy wall filter flask with a tooled neck for uniform stopper fit. Side hose connection is a #11 Ace-Thred that accepts a PTFE tubing connector with a silicone o-ring and nylon bushing to make an easier, safer, more convenient vacuum hose connection. Includes flask, 3/8 tubing connector, and nylon bushing.