Learn More
Chemglass Life Sciences 250mL Erlenmeyer Flask
SDP
Catalog No. 501215021
Encompass_Preferred
250mL Erlenmeyer Flask. Tops are reinforced and tooled with a rounded finish, body is thick walled with a long tapered outside contour to minimize chipping when struck or rubbed together. All flasks have durable white ceramic enamel scales to indicate approximate volumes. Designed from ASTM Specification E1404, Type I, Class 1 requirements.
Catalog No. 50-121-5021 Supplier Chemglass Life Sciences Supplier No. CG8496P250
250mL Erlenmeyer Flask. Tops are reinforced and tooled with a rounded finish, body is thick walled with a long tapered outside contour to minimize chipping when struck or rubbed together. All flasks have durable white ceramic enamel scales to indicate approximate volumes. Designed from ASTM Specification E1404, Type I, Class 1 requirements.