Learn More
Chemglass Life Sciences 250mL Erlenmeyer Flask, Iodine, Stopper, COMPLETE
SDP
Catalog No. CG155204
Encompass_Preferred
250mL Erlenmeyer Flask, Iodine, Stopper, COMPLETE. Flask has a funnel top with a standard taper outer grind to accept a 24/40 long neck stopper allowing the handle to extend above the liquid seal. Used in ASTM D29 and ASTM D555. Available complete with stopper or flask only.
Catalog No. CG155204 Supplier Chemglass Life Sciences Supplier No. CG155204
250mL Erlenmeyer Flask, Iodine, Stopper, COMPLETE. Flask has a funnel top with a standard taper outer grind to accept a 24/40 long neck stopper allowing the handle to extend above the liquid seal. Used in ASTM D29 and ASTM D555. Available complete with stopper or flask only.