Learn More
Thermco Products Inc Thermal Buffer Bottles, 250 mL
Catalog No. NC1729150
Encompass_Preferred
Suspending a sensor probe in a bottle filled with a thermal buffer, such as glycol, will provide more accurate readings of temperatures by averaging out rapid temperature fluctuations from intermittent air currents.
Catalog No. NC1729150 Supplier Thermco Products Inc Supplier No. ACCBOT250BIO
Suspending a sensor probe in a bottle filled with a thermal buffer, such as glycol, will provide more accurate readings of temperatures by averaging out rapid temperature fluctuations from intermittent air currents.