Learn More
CELLTREAT Scientific Products 250mL Graduated Beaker With Handle, Polypropylene, Non-sterile, Case Quantity 12
SDP
Catalog No. 5000670047
Encompass_Preferred
Molded pouring spouts and rolled lip fordripless work environment
Handled Beakers feature clearly printedgraduations
Sturdy handles with textured thumb restensure safe handling of hazardous solutions
Polypropylene material is resistant to avariety of strong acids, bases, and solvents
Can withstand -20 ̊C to 135 ̊C
Catalog No. 50-006-70047 Supplier CELLTREAT Scientific Products Supplier No. 230522
Molded pouring spouts and rolled lip fordripless work environment
Handled Beakers feature clearly printedgraduations
Sturdy handles with textured thumb restensure safe handling of hazardous solutions
Polypropylene material is resistant to avariety of strong acids, bases, and solvents
Can withstand -20 ̊C to 135 ̊C