Learn More
Ace Glass, Inc. 250mL hemispherical fabric mantle, Glas-Col, 83mm max flask diameter, 1.89in inside depth, 4.77in OD, 2.64in height, 180 watts, CSA rated
SDP
Catalog No. 509558953
Encompass
250mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.
Catalog No. 50-955-8953 Supplier Ace Glass, Inc. Supplier No. 1203513
250mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.