Learn More
Chemglass Life Sciences 250mL Single Neck Round Bottom Flask (RBF) , 14/20 Outer with Hooks
SDP
Catalog No. CG150713
Encompass_Preferred
250mL Single Neck Round Bottomed Flask, 14/20 Outer with Hooks. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint. Flask has two hooks 180¿ apart for use with CG-110 stainless steel springs.
Catalog No. CG150713 Supplier Chemglass Life Sciences Supplier No. CG150713
250mL Single Neck Round Bottomed Flask, 14/20 Outer with Hooks. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint. Flask has two hooks 180¿ apart for use with CG-110 stainless steel springs.