Learn More
Ace Glass, Inc. 250mL separatory funnel with locking stopper
SDP
Catalog No. 50979221
Encompass_Preferred
250ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-979-221 Supplier Ace Glass, Inc. Supplier No. 723159
250ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.