Learn More
CELLTREAT Scientific Products 250mL Media Bottle, Square, PET, Individually Wrapped, Sterile
SDP
Catalog No. NC2206462
Encompass_Preferred
Lightweight plastic material is easier and safer to handle than glass. Square bottle shape conserves space in refrigerated storage areas. Wide mouth design minimizes contamination risk when pipetting media and solutions. Clearly printed graduations for quick volume reference. Defined marking area is easy to write on. Bottles are sterile, available individually wrapped or tray packed with 12 bottles per tray. PET material reduces gas permeability and improves solution stability compared to polystyrene. Both PET and PETG media bottles have equivalent performance and share key material properties, such as operating temperature range (-80°C to 60°C) and chemical compatibility
This item is not returnable. View return policy
Catalog No. NC2206462 Supplier CELLTREAT Scientific Products Supplier No. 229789
Lightweight plastic material is easier and safer to handle than glass. Square bottle shape conserves space in refrigerated storage areas. Wide mouth design minimizes contamination risk when pipetting media and solutions. Clearly printed graduations for quick volume reference. Defined marking area is easy to write on. Bottles are sterile, available individually wrapped or tray packed with 12 bottles per tray. PET material reduces gas permeability and improves solution stability compared to polystyrene. Both PET and PETG media bottles have equivalent performance and share key material properties, such as operating temperature range (-80°C to 60°C) and chemical compatibility