Learn More
Ace Glass, Inc. 250mL separatory funnel, pear shaped, complete with #11removable PTFE drip tip and 2mm bore PTFE stopcock plug, 12632-22 size 22 PTFE stopper not included
SDP
Catalog No. 50974670
Encompass_Preferred
250ml pear-shaped, separatory funnel complete with #11 removable PTFE drip tip and 2mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.
Catalog No. 50-974-670 Supplier Ace Glass, Inc. Supplier No. 722353
250ml pear-shaped, separatory funnel complete with #11 removable PTFE drip tip and 2mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.