Learn More
Environmental Sampling Supply Inc ESS 250 mL HDPE Oblong Bottle, Leakproof, Graduated, 250 mL
Catalog No. NC1405338
Encompass_Preferred
The ESS 250ML CapSure™ bottle is crafted from high-density polyethylene (HDPE) and features a reliable leak-tight closure system, perfect for safe environmental and analytical sample storage. Its wide-mouth design allows for easy filling and dispensing, while being compliant with U.S. EPA cleaning standards. This durable and chemical-resistant container is designed for professional use, making it a preferred choice for labs requiring high-quality, certified sample containment.
Catalog No. NC1405338 Supplier Environmental Sampling Supply Inc Supplier No. 02501902QC
The ESS 250ML CapSure™ bottle is crafted from high-density polyethylene (HDPE) and features a reliable leak-tight closure system, perfect for safe environmental and analytical sample storage. Its wide-mouth design allows for easy filling and dispensing, while being compliant with U.S. EPA cleaning standards. This durable and chemical-resistant container is designed for professional use, making it a preferred choice for labs requiring high-quality, certified sample containment.