Learn More
CELLTREAT Scientific Products 250mL Centrifuge Bottles, Polycarbonate, Knurled Seal Cap, Non-sterile
SDP
Catalog No. 501656690
Encompass_Preferred
Polycarbonate Centrifuge Bottles have a rigid body for moderate speed centrifugation of biological and chemical samples. Seal cap has extended lip to facilitate removal from rotor, and gripping ridges for easy opening and sealing. Bottles include molded graduation lines. Bottle must be filled 80% of total capacity to obtain maximum results. High quality polycarbonate has excellent chemical resistance. Reusable and autoclavable. Packaged non-sterile in re-sealable bags. Use of a rotor cushion is required to support botthes during centrifugation. Made in the USA. Every case is printed with the lot number for quality traceability. Packed non-sterile, 4/case.
Catalog No. 50-165-6690 Supplier CELLTREAT Scientific Products Supplier No. 229464
Polycarbonate Centrifuge Bottles have a rigid body for moderate speed centrifugation of biological and chemical samples. Seal cap has extended lip to facilitate removal from rotor, and gripping ridges for easy opening and sealing. Bottles include molded graduation lines. Bottle must be filled 80% of total capacity to obtain maximum results. High quality polycarbonate has excellent chemical resistance. Reusable and autoclavable. Packaged non-sterile in re-sealable bags. Use of a rotor cushion is required to support botthes during centrifugation. Made in the USA. Every case is printed with the lot number for quality traceability. Packed non-sterile, 4/case.