Learn More
Chemglass Life Sciences 250mL Pear Shaped Flask, 29/42 Outer Joint
SDP
Catalog No. 501215247
Encompass_Preferred
250mL Pear Shaped Flask, 29/42 Outer Joint. Pear shape HEAVY WALL flask with a standard taper outer joint.
Catalog No. 50-121-5247 Supplier Chemglass Life Sciences Supplier No. CG155405
250mL Pear Shaped Flask, 29/42 Outer Joint. Pear shape HEAVY WALL flask with a standard taper outer joint.