Learn More
Chemglass Life Sciences 250mL Glassblowers Pear Shaped Blank
SDP
Catalog No. CG62105
Encompass_Preferred
250mL Glassblowers Pear Shaped Blank. Blanks for fabrication of pear-shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. CG62105 Supplier Chemglass Life Sciences Supplier No. CG62105
250mL Glassblowers Pear Shaped Blank. Blanks for fabrication of pear-shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.