Learn More
Chemglass Life Sciences 250mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 32 Thread
SDP
Catalog No. CG186005
Encompass_Preferred
250mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 32 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower PTFE stopcock has a 2mm bore with the arm being 8mm O.D. Vessels are available in sizes other than those listed. Please call for details.
Catalog No. CG186005 Supplier Chemglass Life Sciences Supplier No. CG186005
250mL Peptide Synthesis Vessel, Solid Phase, Medium Frit, GL 32 Thread. Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support. Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Lower PTFE stopcock has a 2mm bore with the arm being 8mm O.D. Vessels are available in sizes other than those listed. Please call for details.