Learn More
Chemglass Life Sciences 250mL Peptide Synthesis Reaction Vessel, Jacketed, Lower Drain, Coarse Frit, 60mm ID Flange, Drain Valve Type/CG-511-01, 60mm Body I.D. x 110mm Height
SDP
Catalog No. CG186802
Encompass_Preferred
Glass Vessel Only, Supplied Complete with (2) CG-4910-15 Caps, Bill-Board. Vessels are 75mm long with 13-425 Threads on either end. A fritted support is sealed into the vessel. Supplied with 2 caps. Part of the CG-1869-01 Bill-Board™ set. The Bill-Board™ set has been designed to simplify and expedite multiple, solid-phase syntheses. Up to 6 reactions can be conducted simultaneously in fritted glass reaction vessels. The Bill-Board™ equipment keeps the solid-phase reactions organized in a grid (for potential combinatorial reaction sequences). Also, the system simplifies the essential steps of solid-phase synthesis which involve repeated cycles of reactions and washings, followed by a final cleavage, product collection and solvent evaporation. The Bill-Boards can also be used for any procedures that require multiple filtrations (i.e. simultaneous silica gel chromatography).
Catalog No. CG186802 Supplier Chemglass Life Sciences Supplier No. CG186802
Glass Vessel Only, Supplied Complete with (2) CG-4910-15 Caps, Bill-Board. Vessels are 75mm long with 13-425 Threads on either end. A fritted support is sealed into the vessel. Supplied with 2 caps. Part of the CG-1869-01 Bill-Board™ set. The Bill-Board™ set has been designed to simplify and expedite multiple, solid-phase syntheses. Up to 6 reactions can be conducted simultaneously in fritted glass reaction vessels. The Bill-Board™ equipment keeps the solid-phase reactions organized in a grid (for potential combinatorial reaction sequences). Also, the system simplifies the essential steps of solid-phase synthesis which involve repeated cycles of reactions and washings, followed by a final cleavage, product collection and solvent evaporation. The Bill-Boards can also be used for any procedures that require multiple filtrations (i.e. simultaneous silica gel chromatography).