Learn More
Alkali Scientific 250mL PETG Square Storage Bott
SDP
Supplier:  Alkali Scientific EFC804
Encompass_Preferred
The Liquid Transfer System collection provides an aseptic transfer solution of closed liquid infusion for the pharmaceuticals and biotechnology industry as well as laboratories A standard Luer head is used in the joint of the system enabling the operation easier and more convenient Also the TPE material of tubes is in line with the requirements for industrial pharmaceutical enterprises and is validated by a certified third party in accordance with BPOG Guide for Evaluating Extractables and Leachables Complete piping specifications Transfer caps with integrated injection molding wide versatility can be adapted to other brands PETG wall is thick durable and resistant to fracture High chemical durability suitable for the shortage of high purity reagent standard particle materials and samples High transparency strong mechanical strength and shock resistance and convenient for observation and transportation Highly tolerant of low temperature UV rays and resistant to fracture Smooth i
Catalog No. 50-302-7564