Learn More
Ace Glass, Inc. Photochem reactor body, 250mL, complete with spinbar, sparger, bushings and PTFE tubing
SDP
Catalog No. NC0683575
Encompass_Preferred
250mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Supplied with spinbar, sparger, bushings and PTFE tubing. Utilizes a height-adjustable non-freezing #50 Ace-Thred.
Catalog No. NC0683575 Supplier Ace Glass, Inc. Supplier No. 786336
250mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Supplied with spinbar, sparger, bushings and PTFE tubing. Utilizes a height-adjustable non-freezing #50 Ace-Thred.