Learn More
Ace Glass, Inc. 250mL gas washing bottle with porosity A (145-174 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
SDP
Catalog No. 50979119
Encompass_Preferred
250mL gas washing bottle with porosity A (145-174 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-979-119 Supplier Ace Glass, Inc. Supplier No. 716660
250mL gas washing bottle with porosity A (145-174 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.