Learn More
Ace Glass, Inc. 250mL complete gas washing bottle, porosity A
SDP
Supplier:  Ace Glass, Inc. 716212
Encompass_Preferred
250mL complete gas washing bottle with a 30mm fritted porosity A (145-174micron) disc.
Catalog No. 50-979-105