Learn More
Ace Glass, Inc. 250mL complete gas washing bottle, porosity A
SDP
Supplier:  Ace Glass, Inc. 716312
Encompass_Preferred
250mL gas washing bottle with porosity A (145-174 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-979-110