Learn More
Ace Glass, Inc. 250mL gas wash stopper only, porosity A
SDP
Supplier:  Ace Glass, Inc. 716370
Encompass_Preferred
250mL gas washing stopper with porosity A (145-174 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-979-112