Learn More
Ace Glass, Inc. 250mL gas wash stopper only, porosity A
SDP
Catalog No. 50974105
Encompass_Preferred
250mL gas wash stopper with porosity A (145-174micron) fritted disc.
Catalog No. 50-974-105 Supplier Ace Glass, Inc. Supplier No. 716270
250mL gas wash stopper with porosity A (145-174micron) fritted disc.