Learn More
Ace Glass, Inc. 250mL gas washing bottle with porosity B (70-100 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
SDP
Catalog No. 50974139
Encompass_Preferred
250mL gas washing bottle with porosity B (70-100 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-139 Supplier Ace Glass, Inc. Supplier No. 716662
250mL gas washing bottle with porosity B (70-100 micron) fritted disc and 29/42 joint. Inlet/outlet arms are 8mm O.D.