Learn More
Ace Glass, Inc. 250mL complete gas washing bottle, porosity B
SDP
Catalog No. 50974112
Encompass_Preferred
250mL gas washing bottle with porosity B (70-100 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-112 Supplier Ace Glass, Inc. Supplier No. 716314
250mL gas washing bottle with porosity B (70-100 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.