Learn More
Ace Glass, Inc. 250mL gas wash stopper only, porosity B
SDP
Catalog No. 50979108
Encompass_Preferred
250mL gas wash stopper with porosity B (70-100micron) fritted disc.
Catalog No. 50-979-108 Supplier Ace Glass, Inc. Supplier No. 716272
250mL gas wash stopper with porosity B (70-100micron) fritted disc.