Learn More
Ace Glass, Inc. 250mL gas wash stopper only, porosity B
SDP
Supplier:  Ace Glass, Inc. 716372
Encompass_Preferred
250mL gas washing stopper with porosity B (70-100 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-118