Learn More
Ace Glass, Inc. 250mL gas wash stopper only, porosity C
SDP
Catalog No. 50974106
Encompass_Preferred
250mL gas wash stopper with porosity C (25-50micron) fritted disc.
Catalog No. 50-974-106 Supplier Ace Glass, Inc. Supplier No. 716274
250mL gas wash stopper with porosity C (25-50micron) fritted disc.