Learn More
Ace Glass, Inc. 250ML SIDE POWDER DISP FUNNEL
SDP
Catalog No. 50974720
Encompass_Preferred
250mL side-operated, screw-feed powder dispensing funnel, for use with up to 25 mesh. Features a #15 Ace-Thred at the top, a large, 1-1/4” knob, and top female and bottom male 24/40 joints.
Catalog No. 50-974-720 Supplier Ace Glass, Inc. Supplier No. 723940
250mL side-operated, screw-feed powder dispensing funnel, for use with up to 25 mesh. Features a #15 Ace-Thred at the top, a large, 1-1/4” knob, and top female and bottom male 24/40 joints.