Learn More
Chemglass Life Sciences 250mL Powder Addition Funnel, 24/40 Joint Size, Auger B
SDP
Catalog No. CG172313
Encompass_Preferred
250mL Powder Addition Funnel, 24/40 Joint Size, Auger B. Facilitates the delivery of free-flowing, granular solids into reactions. Pressure equalizing arm is equipped with a vented stopper for air-free operations and also permits access into the delivery stem for easy clean-out. One piece, easily removed grooved PTFE auger delivers powders at a uniform rate. The 250 and 500mL capacities have the larger, deep cut auger (Size B) for use with coarse material. Funnel has a lower inner joint and top outer, both being the size listed.
Catalog No. CG172313 Supplier Chemglass Life Sciences Supplier No. CG172313
250mL Powder Addition Funnel, 24/40 Joint Size, Auger B. Facilitates the delivery of free-flowing, granular solids into reactions. Pressure equalizing arm is equipped with a vented stopper for air-free operations and also permits access into the delivery stem for easy clean-out. One piece, easily removed grooved PTFE auger delivers powders at a uniform rate. The 250 and 500mL capacities have the larger, deep cut auger (Size B) for use with coarse material. Funnel has a lower inner joint and top outer, both being the size listed.