Learn More
MAGIC BIOPROCESSING MAGic™Beads Protein G 250mL
Catalog No. NC3723549
Encompass_Preferred
MAGic™ Beads Protein G are super-paramagnetic porous beads coupled with a Protein G ligand, optimized for antibodies that lack affinity for Protein A. Purify directly from hybridoma or CHO cell harvests while bypassing centrifugation and filtration. MAGic™ Beads Protein G offer high IgG binding capacity and strong magnetic responsiveness, enabling rapid and efficient separation even in large-volume samples. This makes them ideal for high-throughput, scalable antibody purification workflows where speed, reliability, and ease of use are essential. Supplied as a convenient 10% slurry, MAGic™ Beads Protein G deliver fast, efficient purification for a broader range of antibodies.
Catalog No. NC3723549 Supplier MAGIC BIOPROCESSING Supplier No. 1425
MAGic™ Beads Protein G are super-paramagnetic porous beads coupled with a Protein G ligand, optimized for antibodies that lack affinity for Protein A. Purify directly from hybridoma or CHO cell harvests while bypassing centrifugation and filtration. MAGic™ Beads Protein G offer high IgG binding capacity and strong magnetic responsiveness, enabling rapid and efficient separation even in large-volume samples. This makes them ideal for high-throughput, scalable antibody purification workflows where speed, reliability, and ease of use are essential. Supplied as a convenient 10% slurry, MAGic™ Beads Protein G deliver fast, efficient purification for a broader range of antibodies.