Learn More
Chemglass Life Sciences Flask, Round Bottom, 250mL, Heavy Wall, 3-Neck, 29/26 CN - 29/26 SN's Angled 20°
SDP
Catalog No. CG152490
Encompass_Preferred
Flask, Round Bottom, 250mL, Heavy Wall, 29/26 - 29/26, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/26Side neck: 29/26
Catalog No. CG152490 Supplier Chemglass Life Sciences Supplier No. CG152490
Flask, Round Bottom, 250mL, Heavy Wall, 29/26 - 29/26, 3-Neck, Angled 20¿. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and 20¿ angled side necks.Center neck: 29/26Side neck: 29/26