Learn More
Chemglass Life Sciences 250mL 2-Neck Round Bottom Flask (RBF) , 1-CN 24/40 Outer, 1-SN 24/40 Outer
SDP
Catalog No. CG151803
Encompass_Preferred
250mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.
Catalog No. CG151803 Supplier Chemglass Life Sciences Supplier No. CG151803
250mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.