Learn More
Chemglass Life Sciences 250mL Reaction Vessel, Jacketed, Graduated Both Sides, 100mm Flange, Flat Flange Inlet/Outlet, CG-911-A Drain Valve
SDP
Catalog No. 5000980636
Encompass_Preferred
Features:. Tool-Free Jacket Connection Design Allows Hose to be Attached or Removed by Hand Without Using Tools. . Spring Loaded Drain Valve for Temperature Applications From -60 C to 230 C with Perfluoro Class VI O-Rings. . Graduation Scale on Both Sides Showing the Approximate Volume. . Sidearm on Lower Drain Valve can be Used with a Standard 3/4” TFE, PFA or Polypropylene Compression Fitting. . All Vessels are Serial Numbered. . Jacketed reaction vessel with graduation scale on both sides. Inlet/outlet hastool-free flat flange connector. No Tools Are Necessary to attach hose fittings to the inlet and outlet of the jacket. Vessels are available with0-25mm spring loaded drain valve supplied complete with one-piece plug and control knob for temperature applications from -60 C to 230 C. Spring loaded drain valves compensate for temperature related material changes and are designed to provide a constant leak-free seal. The sidearm on the valve is 3/4” Swagelok. Valves are designed to sea
Catalog No. 50-009-80636 Supplier Chemglass Life Sciences Supplier No. CG1929X09
Features:. Tool-Free Jacket Connection Design Allows Hose to be Attached or Removed by Hand Without Using Tools. . Spring Loaded Drain Valve for Temperature Applications From -60 C to 230 C with Perfluoro Class VI O-Rings. . Graduation Scale on Both Sides Showing the Approximate Volume. . Sidearm on Lower Drain Valve can be Used with a Standard 3/4” TFE, PFA or Polypropylene Compression Fitting. . All Vessels are Serial Numbered. . Jacketed reaction vessel with graduation scale on both sides. Inlet/outlet hastool-free flat flange connector. No Tools Are Necessary to attach hose fittings to the inlet and outlet of the jacket. Vessels are available with0-25mm spring loaded drain valve supplied complete with one-piece plug and control knob for temperature applications from -60 C to 230 C. Spring loaded drain valves compensate for temperature related material changes and are designed to provide a constant leak-free seal. The sidearm on the valve is 3/4” Swagelok. Valves are designed to sea